Terephthaldehyde monodiethylacetal, 97%, stabilized
Terephthaldehyde monodiethylacetal, 97%, stabilized
Terephthaldehyde monodiethylacetal, 97%, stabilized
Terephthaldehyde monodiethylacetal, 97%, stabilized
Thermo Scientific Chemicals
Terephthaldehyde monodiethylacetal, 97%, stabilized
CAS: 81172-89-6 | C12H16O3 | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA294630250
Price (MYR)
807.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
807.00
EA
Add to cart
Specifications
Chemical Name or MaterialTerephthaldehyde monodiethylacetal
Name Note97%
CAS81172-89-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: