3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals
3,4-Dichlorobenzaldehyde, 98%
CAS: 6287-38-3 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA29456025025 gGlass bottle
Catalog number FSA294560250
Price (MYR)
456.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-Dichlorobenzaldehyde
Name Note98%
CAS6287-38-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger