Bis(diphenylphosphino)methane, 98%
Bis(diphenylphosphino)methane, 98%
Bis(diphenylphosphino)methane, 98%
Bis(diphenylphosphino)methane, 98%
Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%
CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA293610250
Price (MYR)
1,587.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,587.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(diphenylphosphino)methane
Name Note98%
CAS2071-20-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning