Check Order Status
4-(Trifluoromethoxy)benzyl bromide, 98%, Thermo Scientific™
Element Search
Structure Search
4-(Trifluoromethoxy)benzyl bromide, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number
FSA293510025
Price (MYR)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
4-(Trifluoromethoxy)benzyl bromide
CAS
50824-05-0
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Danger
CAS Max %
100.0