Cobalt(II) acetate tetrahydrate, 98+%
Cobalt(II) acetate tetrahydrate, 98+%
Cobalt(II) acetate tetrahydrate, 98+%
Cobalt(II) acetate tetrahydrate, 98+%
Thermo Scientific Chemicals
Cobalt(II) acetate tetrahydrate, 98+%
CAS: 6147-53-1 | C4H6CoO4·4H2O | 249.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA293062500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II) acetate tetrahydrate
Melting Point140.0°C
Name Note97%
CAS6147-53-1
ColorPink to Pink-Red