o-Tolyl isocyanate, 99+%
o-Tolyl isocyanate, 99+%
o-Tolyl isocyanate, 99+%
o-Tolyl isocyanate, 99+%
o-Tolyl isocyanate, 99+%
CAS: 614-68-6 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA292220250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialo-Tolyl isocyanate
Name Note99+%
CAS614-68-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.