Isobornyl methacrylate, 85-90%, stabilized
Isobornyl methacrylate, 85-90%, stabilized
Isobornyl methacrylate, 85-90%, stabilized
Isobornyl methacrylate, 85-90%, stabilized
Thermo Scientific Chemicals
Isobornyl methacrylate, 85-90%, stabilized
CAS: 7534-94-3 | C14H22O2 | 222.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA292090010
Price (MYR)
1,301.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,301.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsobornyl methacrylate
Name Note85-90%
CAS7534-94-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.