2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
2-Chloro-1H-benzimidazole, 97%
CAS: 4857-06-1 | C7H5ClN2 | 152.58 g/mol
Have Questions?
Catalog number FSA291960250
Price (MYR)
1,225.00
G
Add to cart
Request bulk or custom format
Price (MYR)
1,225.00
G
Add to cart
Specifications
Chemical Name or Material2-Chloro-1H-benzimidazole
CAS4857-06-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0