Dicobalt octacarbonyl, 95%, stabilized
Dicobalt octacarbonyl, 95%, stabilized
Dicobalt octacarbonyl, 95%, stabilized
Dicobalt octacarbonyl, 95%, stabilized
Thermo Scientific Chemicals
Dicobalt octacarbonyl, 95%, stabilized
CAS: 10210-68-1 | C8Co2O8 | 341.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA291840250
Price (MYR)
2,187.00
G
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,187.00
G
Add to cart
Specifications
Chemical Name or MaterialDicobalt octacarbonyl
Name Note95%
CAS10210-68-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Self-heating; may catch fire.