(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA291830010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material(±)-Miconazole nitrate salt
Type(±)-Miconazole Nitrate Salt
CAS22832-87-7
Health Hazard 1Warning
Health Hazard 2GHS H Statement Harmful if swallowed. May cause an allergic skin reaction.