Potassium sorbate, 99%
Potassium sorbate, 99%
Potassium sorbate, 99%
Potassium sorbate, 99%
Thermo Scientific Chemicals
Potassium sorbate, 99%
CAS: 24634-61-5 | C6H7KO2 | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA291742500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium sorbate
AcidityPasses test
CAS24634-61-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement