Calix[4]arene, 98%
Calix[4]arene, 98%
Calix[4]arene, 98%
Thermo Scientific Chemicals
Calix[4]arene, 98%
CAS: 74568-07-3 | C28H24O4 | 424.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA290190050
Price (MYR)
3,729.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,729.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalix[4]arene
Name Note98%
CAS74568-07-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
View more
Chelation/Complexation reagent
RUO – Research Use Only