Ursolic acid, 95%
Ursolic acid, 95%
Ursolic acid, 95%
Thermo Scientific Chemicals
Ursolic acid, 95%
CAS: 77-52-1 | C30H48O3 | 456.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Packaging:
Glass bottle
Catalog number FSA290182500
Price (MYR)
1,088.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,088.00
BOT
Add to cart
Specifications
Chemical Name or MaterialUrsolic acid
CAS77-52-1
Melting Point283.0°C to 285.0°C
ColorWhite
EINECS Number201-034-