Ruthenium Red, ≥85%, pure
Ruthenium Red, ≥85%, pure
Ruthenium Red, ≥85%, pure
Ruthenium Red, ≥85%, pure
Thermo Scientific Chemicals
Ruthenium Red, ≥85%, pure
CAS: 11103-72-3 | H24Cl6N14O2Ru3 | 786.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA279710010
Price (MYR)
1,133.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,133.00
EA
Add to cart
Specifications
Chemical Name or MaterialRuthenium Red
Melting Point> 500.0°C
CAS11103-72-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement