2-Aminobenzaldehyde, 97%
2-Aminobenzaldehyde, 97%
2-Aminobenzaldehyde, 97%
2-Aminobenzaldehyde, 97%
Thermo Scientific Chemicals
2-Aminobenzaldehyde, 97%
CAS: 529-23-7 | C7H7NO | 121.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2796600101 gGlass bottle
Catalog number FSA279660010
Price (MYR)
1,067.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,067.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Aminobenzaldehyde
CAS529-23-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0