N,N-Dimethylformamide, 99.5%, for HPLC
N,N-Dimethylformamide, 99.5%, for HPLC
N,N-Dimethylformamide, 99.5%, for HPLC
N,N-Dimethylformamide, 99.5%, for HPLC
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for HPLC
For HPLC | CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA279600010
Price (MYR)
678.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
678.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 350nm, 0.015 max. at 340nm, 0.02 max. at 320nm, 0.05 max. at 300nm, 0.15 max. at 280nm, 0.5 max. at 270nm
CAS68-12-2
CAS Min %99.4
View more
For HPLC
RUO – Research Use Only