4'-Iodoacetophenone, 98%
4'-Iodoacetophenone, 98%
4'-Iodoacetophenone, 98%
4'-Iodoacetophenone, 98%
Thermo Scientific Chemicals
4'-Iodoacetophenone, 98%
CAS: 13329-40-3 | C8H7IO | 246.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA278890250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4'-Iodoacetophenone
Name Note98%
CAS13329-40-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning