Brucine, 99%, anhydrous
Brucine, 99%, anhydrous
Brucine, 99%, anhydrous
Brucine, 99%, anhydrous
Thermo Scientific Chemicals
Brucine, 99%, anhydrous
CAS: 357-57-3 | C23H26N2O4 | 394.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA278720050
Price (MYR)
289.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
289.00
EA
Add to cart
Specifications
Chemical Name or MaterialBrucine
Name NoteAnhydrous
CAS357-57-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Fatal if swallowed.
Harmful to aquatic life with long lasting effects.
Fatal if inhaled.