Polysorbate 80
Polysorbate 80
Polysorbate 80
Thermo Scientific Chemicals
Polysorbate 80
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Catalog number FSA278632500
Price (MYR)
288.00
BOT
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
288.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPolysorbate 80
CAS9005-65-6
ColorYellow to Amber
Density1.0800 g/mL
Quantity250 mL
View more
RUO – Research Use Only
Contents & Storage
Indoors +20°C