Triacontane, 98%
Triacontane, 98%
Triacontane, 98%
Thermo Scientific Chemicals
Triacontane, 98%
CAS: 638-68-6 | C30H62 | 422.826 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA278050010
Price (MYR)
514.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
514.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriacontane
CAS638-68-6
Melting Point65°C to 67°C
ColorWhite
EINECS Number211-349-5