2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals
CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA278031000
Price (MYR)
1,773.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,773.00
EA
Add to cart
Specifications
Chemical Name or Material2-Aminoterephthalic acid
CAS10312-55-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0