4,4'-Dipyridyl, 98%
4,4'-Dipyridyl, 98%
4,4'-Dipyridyl, 98%
Thermo Scientific Chemicals
4,4'-Dipyridyl, 98%
CAS: 553-26-4 | C10H8N2 | 156.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA276200250
Price (MYR)
618.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
618.00
BOT
Add to cart
Specifications
Chemical Name or Material4, 4'-Dipyridyl
CAS553-26-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist