Water, pure, demineralized
Water, pure, demineralized
Water, pure, demineralized
Water, pure, demineralized
Thermo Scientific Chemicals
Water, pure, demineralized
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
5 L
Packaging:
View all
Plastic Can
Catalog number FSA276010200
Price (MYR)
1,155.00
LT
Add to cart
Quantity:
20 L
Packaging:
Plastic Can
Request bulk or custom format
Price (MYR)
1,155.00
LT
Add to cart
Specifications
Chemical Name or MaterialWater
pH6 to 8
CAS7732-18-5
ColorWhite
Health Hazard 1
View more
Biological buffer
RUO – Research Use Only