Tripropionin, 97%
Tripropionin, 97%
Tripropionin, 97%
Thermo Scientific Chemicals
Tripropionin, 97%
CAS: 139-45-7 | C12H20O6 | 260.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA275101000
Price (MYR)
361.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
361.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTripropionin
Name Note97%
CAS139-45-7
Health Hazard 1LIGHT SENSITIVE