Di-2-ethylhexylamine, 99%
Di-2-ethylhexylamine, 99%
Di-2-ethylhexylamine, 99%
Di-2-ethylhexylamine, 99%
Thermo Scientific Chemicals
Di-2-ethylhexylamine, 99%
CAS: 106-20-7 | C16H35N | 241.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number FSA271962500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-2-ethylhexylamine
Name Note99%
CAS106-20-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: