2-Ethylhexylamine, 99%
2-Ethylhexylamine, 99%
2-Ethylhexylamine, 99%
2-Ethylhexylamine, 99%
Thermo Scientific Chemicals
2-Ethylhexylamine, 99%
CAS: 104-75-6 | C8H19N | 129.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA271660010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethylhexylamine
CAS104-75-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0