9-Hydroxyxanthene, 97+%
9-Hydroxyxanthene, 97+%
9-Hydroxyxanthene, 97+%
9-Hydroxyxanthene, 97+%
Thermo Scientific Chemicals
9-Hydroxyxanthene, 97+%
CAS: 90-46-0 | C13H10O2 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA271520050
Price (MYR)
452.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
452.00
BOT
Add to cart
Specifications
Chemical Name or Material9-Hydroxyxanthene
Name Note99%
CAS90-46-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning