2,4-Difluorobenzaldehyde, 98%
2,4-Difluorobenzaldehyde, 98%
2,4-Difluorobenzaldehyde, 98%
2,4-Difluorobenzaldehyde, 98%
Thermo Scientific Chemicals
2,4-Difluorobenzaldehyde, 98%
CAS: 1550-35-2 | C7H4F2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA271470250
Price (MYR)
1,142.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,142.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Difluorobenzaldehyde
CAS1550-35-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0