2,4-Dimethylstyrene, 97%, stabilized
2,4-Dimethylstyrene, 97%, stabilized
2,4-Dimethylstyrene, 97%, stabilized
2,4-Dimethylstyrene, 97%, stabilized
Thermo Scientific Chemicals
2,4-Dimethylstyrene, 97%, stabilized
CAS: 2234-20-0 | C10H12 | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA271150050
Price (MYR)
514.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
514.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Dimethylstyrene
Name Note97%
CAS2234-20-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning