delta-Gluconolactone, 99%
delta-Gluconolactone, 99%
delta-Gluconolactone, 99%
delta-Gluconolactone, 99%
Thermo Scientific Chemicals
delta-Gluconolactone, 99%
CAS: 90-80-2 | C6H10O6 | 178.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA271052500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialδ-Gluconolactone
CAS90-80-2
Health Hazard 1LIGHT SENSITIVE
Health Hazard 2MOISTURE SENSITIVE
Melting Point151.0°C to 155.0°C