Fehling A solution
Fehling A solution
Fehling A solution
Thermo Scientific Chemicals
Fehling A solution
CAS: 7758-98-7 | CuO4S | 159.602 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number FSA270800025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFehling A solution
ColorBlue
CAS7758-98-7
Molecular FormulaCuO4S
Linear FormulaCuSO4