PYBOP™, 99%
PYBOP™, 99%
PYBOP™, 99%
Thermo Scientific Chemicals
PYBOP™, 99%
CAS: 128625-52-5 | C18H28F6N6OP2 | 520.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA270720050
Price (MYR)
532.00
BOT
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
532.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPYBOP*
Name Note99%
CAS128625-52-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Coupling reagent
RUO – Research Use Only