Alginic acid
Alginic acid
Alginic acid
Thermo Scientific Chemicals
Alginic acid
CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA270585000
Price (MYR)
1,782.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,782.00
EA
Add to cart
Specifications
Chemical Name or MaterialAlginic acid
CAS9005-32-7
Melting Point>300°C
ColorBeige to White
RTECS NumberAZ5775000