Formic acid, 99%, for analysis
Formic acid, 99%, for analysis
Formic acid, 99%, for analysis
Formic acid, 99%, for analysis
Thermo Scientific Chemicals
Formic acid, 99%, for analysis
Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated | CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number FSA270480250
Price (MYR)
290.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
290.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFormic acid
CAS64-18-6
Melting Point8.0°C
Boiling Point101.0°C
Health Hazard 1Danger
View more
For analysis
RUO – Research Use Only