Oleic acid, 97%
Oleic acid, 97%
Oleic acid, 97%
Thermo Scientific Chemicals
Oleic acid, 97%
CAS: 112-80-1 | C18H34O2 | 282.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA270291000
Price (MYR)
4,866.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,866.00
EA
Add to cart
Specifications
Chemical Name or MaterialOleic acid
CAS112-80-1
Health Hazard 1Warning
Health Hazard 2GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation.
Health Hazard 3GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.