Perfluorooctyl iodide, 98%
Perfluorooctyl iodide, 98%
Perfluorooctyl iodide, 98%
Perfluorooctyl iodide, 98%
Thermo Scientific Chemicals
Perfluorooctyl iodide, 98%
CAS: 507-63-1 | C8F17I | 545.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA269960100
Price (MYR)
546.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
546.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPerfluorooctyl iodide
Name Note99%
CAS507-63-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: