Monomethyl monopotassium malonate, 98%
Monomethyl monopotassium malonate, 98%
Monomethyl monopotassium malonate, 98%
Monomethyl monopotassium malonate, 98%
Thermo Scientific Chemicals
Monomethyl monopotassium malonate, 98%
CAS: 38330-80-2 | C4H5O4·K | 156.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA269550250
Price (MYR)
318.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMonomethyl monopotassium malonate
CAS38330-80-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0