4-Pentenoic acid, 99%
4-Pentenoic acid, 99%
4-Pentenoic acid, 99%
4-Pentenoic acid, 99%
Thermo Scientific Chemicals
4-Pentenoic acid, 99%
CAS: 591-80-0 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA269020050
Price (MYR)
296.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
296.00
EA
Add to cart
Specifications
Chemical Name or Material4-Pentenoic acid
CAS591-80-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre