Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™
CAS: 4111-54-0 | C6H14LiN | 107.125 g/mol
Have Questions?
Change view
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA268831000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium diisopropylamide
ColorBrown-Red or Orange to Yellow
Name Note2M Solution in THF/n-Heptane
CAS4111-54-0
Boiling Point60.0°C to 136.0°C