5-Sulfosalicylic acid dihydrate, 99+%
5-Sulfosalicylic acid dihydrate, 99+%
5-Sulfosalicylic acid dihydrate, 99+%
5-Sulfosalicylic acid dihydrate, 99+%
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, 99+%
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA268792500
Price (MYR)
369.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
369.00
BOT
Add to cart
Chemical Identifiers
CAS5965-83-3, 97-05-2
Specifications
Chemical Name or Material5-Sulfosalicylic acid dihydrate
Name Notep.a.
CAS5965-83-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
View more
Indicator
RUO – Research Use Only