Lithium acetate, anhydrous, 99%, extra pure
Lithium acetate, anhydrous, 99%, extra pure
Lithium acetate, anhydrous, 99%, extra pure
Lithium acetate, anhydrous, 99%, extra pure
Thermo Scientific Chemicals
Lithium acetate, anhydrous, 99%, extra pure
CAS: 546-89-4 | C2H3LiO2 | 65.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA268640010
Price (MYR)
2,346.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
2,346.00
EA
Add to cart
Specifications
Chemical Name or MaterialLithium acetate, anhydrous
CAS546-89-4
Health Hazard 1GHS Signal Word: Warning
Melting Point280.0°C to 285.0°C
ColorWhite to Yellow