Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Thermo Scientific Chemicals
Hexanes, for HPLC, contains ca. 95% n-Hexane
CAS: 92112-69-1 | C6H14
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA268360025
Price (MYR)
612.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
612.00
BOT
Add to cart
Chemical Identifiers
CAS92112-69-1, 110-54-3
Specifications
Chemical Name or MaterialHexanes
Name Note(95% n-Hexane)
Absorbance(1 cm cell vs HPLC−grade water), 0.005 max. at 250nm, 0.01 max. at 240nm, 0.03 max. at 230nm, 0.07 max. at 220nm, 0.25 max. at 210nm, 0.7 max. at 200nm
CAS92112-69-1
Health Hazard 1Danger
View more
For HPLC
RUO – Research Use Only