Ethyl acetate, 99.5+%, for HPLC
Ethyl acetate, 99.5+%, for HPLC
Ethyl acetate, 99.5+%, for HPLC
Ethyl acetate, 99.5+%, for HPLC
Thermo Scientific Chemicals
Ethyl acetate, 99.5+%, for HPLC
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FSA268350025
Price (MYR)
586.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
586.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 280nm, 0.025 max. at 270nm, 0.12 max. at 260nm, 1 max. at 255nm
CAS141-78-6
ColorColorless
View more
For HPLC
RUO – Research Use Only