Water, for HPLC gradient grade
Water, for HPLC gradient grade
Water, for HPLC gradient grade
Water, for HPLC gradient grade
Thermo Scientific Chemicals
Water, for HPLC gradient grade
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA268300025
Price (MYR)
485.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
485.00
BOT
Add to cart
Specifications
Chemical Name or MaterialWater
Name NoteFor HPLC gradient grade
CAS7732-18-5
ColorColorless
Health Hazard 1
View more
For HPLC
RUO – Research Use Only