Acetonitrile, 99.8%, for HPLC
Acetonitrile, 99.8%, for HPLC
Acetonitrile, 99.8%, for HPLC
Acetonitrile, 99.8%, for HPLC
Thermo Scientific Chemicals
Acetonitrile, 99.8%, for HPLC
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA268270025
Price (MYR)
1,015.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,015.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile
Melting Point-46.0°C
Absorbance(1cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.01 max. at 250nm, 0.015 max. at 240nm, 0.05 max. at 230nm
CAS75-05-8
Health Hazard 1Danger
View more
For HPLC
RUO – Research Use Only