4-Epichlortetracycline hydrochloride, can be used as secondary standard
4-Epichlortetracycline hydrochloride, can be used as secondary standard
4-Epichlortetracycline hydrochloride, can be used as secondary standard
4-Epichlortetracycline hydrochloride, can be used as secondary standard
Thermo Scientific Chemicals
4-Epichlortetracycline hydrochloride, can be used as secondary standard
4-Epichlortetracycline hydrochloride, CAS # 14297-93-9, is a broad-spectrum, bacteriostatic antibiotic of the tetracycline antibiotic family.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA268231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Epichlortetracycline hydrochloride
Name Note97%
CAS101342-45-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
View more
General description
• 4-Epichlortetracycline is the hydrochloride salt of 4-Epichlortetracycline, a tetracycline ring structure with a chloride substitution
• 4-Epichlortetracycline hydrochloride (4-Epi-chlortetracycline) is an epimer of chlortetracycline
• 4-Epichlortetracycline suppresses neutrophil activity and dampens the immune response
• Acts as an aminoacyl-tRNA analog to disrupt polypeptide synthesis, reducing bacterial growth Applications
• Can be used for in vitro research on cancer and inflammatory mechanisms
• Intended for use as a selective agent and secondary standard
RUO – Research Use Only
General References:
  1. Armstrong, A.W.; Hekmatjah, J.; Kircik, L.H. Oral Tetracyclines and Acne: A Systematic Review for Dermatologists. J Drugs Dermatol. 2020, 19(11), s6-s13.
  2. Liou, J.M.; Chen, C.C.; Chang, C.M.; Fang, Y.J.; Bair, M.J.; Chen, P.Y.; Chang, C.Y.; Hsu, Y.C.; Chen, M.J.; Chen, C.C.; Lee, J.Y.; Yang, T.H., Luo, J.C.; Chen, C.Y.; Hsu, W.F.; Chen, Y.N.; Wu, J.Y.; Lin, J.T.; Lu, T.P.; Chuang, E.Y.; El-Omar, E.M.; Wu, M.S. Taiwan Gastrointestinal Disease and Helicobacter Consortium. Long-term changes of gut microbiota, antibiotic resistance, and metabolic parameters after Helicobacter pylori eradication: a multicentre, open-label, randomised trial. Lancet Infect Dis. 2019,(10), 1109-1120.