Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA264780010
Price (MYR)
619.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
619.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMagnesium hydrogen phosphate trihydrate
Name Notec.p., 98%
CAS7782-75-4
ColorWhite
Physical FormPowder