3-Amino-1H-1,2,4-triazole, 95%
3-Amino-1H-1,2,4-triazole, 95%
3-Amino-1H-1,2,4-triazole, 95%
3-Amino-1H-1,2,4-triazole, 95%
Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 95%
CAS: 61-82-5 | C2H4N4 | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA264570100
Price (MYR)
262.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
262.00
EA
Add to cart
Specifications
Chemical Name or Material3-Amino-1H-1, 2, 4-triazole
Name Note95%
CAS61-82-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning