trans-Zeatin (synthetic), 97+%
trans-Zeatin (synthetic), 97+%
trans-Zeatin (synthetic), 97+%
trans-Zeatin (synthetic), 97+%
Thermo Scientific Chemicals
trans-Zeatin (synthetic), 97+%
CAS: 1637-39-4 | C10H13N5O | 219.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
10 mg
Packaging:
Glass bottle
Catalog number FSA264291000
Price (MYR)
2,674.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,674.00
BOT
Add to cart
Specifications
Chemical Name or Materialtrans-Zeatin (synthetic)
Name Note97%
CAS1637-39-4
Melting Point207°C
ColorColorless to Yellow