Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Thermo Scientific Chemicals
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA263820025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzalkonium chloride
ColorColorless to Yellow
CAS8001-54-5
Boiling Point>100.0°C
CAS Min %49.0